missing translation for 'onlineSavingsMsg'
Learn More

1-(Tetrahydro-2-furoyl)piperazine 97.0+%, TCI America™

Catalog No. T26175G
missing translation for 'changeView'
missing translation for 'orderingAttributeHoverText'
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T26175G 5 g
T261725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 missing translation for 'options'
Catalog No. T26175G missing translation for 'mfr' TCI America missing translation for 'supplierNo' T26175G

Chemical Identifiers

CAS 63074-07-7
Molecular Formula C9H16N2O2
Molecular Weight (g/mol) 184.239
MDL Number MFCD00190247
InChI Key UKESBLFBQANJHH-UHFFFAOYSA-N
Synonym 1-tetrahydro-2-furoyl piperazine, 1-2-tetrahydrofuroyl piperazine, piperazin-1-yl tetrahydrofuran-2-yl methanone, 1-2-tetrahydrofuroyl-piperazine, n-2-tetrahydrofuroyl piperazine, 1-oxolane-2-carbonyl piperazine, 1-tetrahydrofuran-2-ylcarbonyl piperazine, n-tetrahydro-2-furoyl piperazine, 1-oxolan-2-ylcarbonyl piperazine, oxolan-2-yl piperazin-1-yl methanone
PubChem CID 2734641
IUPAC Name oxolan-2-yl(piperazin-1-yl)methanone
SMILES C1CC(OC1)C(=O)N2CCNCC2

Specifications

Quantity 5 g
Physical Form Clear Liquid at 20°C
Boiling Point 125°C
Formula Weight 184.24
Percent Purity ≥97.0% (GC)
Chemical Name or Material 1-(Tetrahydro-2-furoyl)piperazine