Learn More
1-(Tetrahydro-2-furoyl)piperazine, 98%
CAS: 63074-07-7 | C9H16N2O2 | 184.24 g/mol
$156.38 - $156.38
Chemical Identifiers
| CAS | 63074-07-7 |
|---|---|
| Molecular Formula | C9H16N2O2 |
| Molecular Weight (g/mol) | 184.24 |
| MDL Number | MFCD00190247 |
| InChI Key | UKESBLFBQANJHH-UHFFFAOYSA-N |
| Synonym | 1-tetrahydro-2-furoyl piperazine, 1-2-tetrahydrofuroyl piperazine, piperazin-1-yl tetrahydrofuran-2-yl methanone, 1-2-tetrahydrofuroyl-piperazine, n-2-tetrahydrofuroyl piperazine, 1-oxolane-2-carbonyl piperazine, 1-tetrahydrofuran-2-ylcarbonyl piperazine, n-tetrahydro-2-furoyl piperazine, 1-oxolan-2-ylcarbonyl piperazine, oxolan-2-yl piperazin-1-yl methanone |
| PubChem CID | 2734641 |
| IUPAC Name | oxolan-2-yl(piperazin-1-yl)methanone |
| SMILES | C1CC(OC1)C(=O)N2CCNCC2 |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
AC367160050
|
Thermo Scientific Chemicals
367160050 |
5 g | Glass bottle |
Each for $156.38
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 63074-07-7 | |
| 184.24 | |
| UKESBLFBQANJHH-UHFFFAOYSA-N | |
| 2734641 | |
| C1CC(OC1)C(=O)N2CCNCC2 |
| C9H16N2O2 | |
| MFCD00190247 | |
| 1-tetrahydro-2-furoyl piperazine, 1-2-tetrahydrofuroyl piperazine, piperazin-1-yl tetrahydrofuran-2-yl methanone, 1-2-tetrahydrofuroyl-piperazine, n-2-tetrahydrofuroyl piperazine, 1-oxolane-2-carbonyl piperazine, 1-tetrahydrofuran-2-ylcarbonyl piperazine, n-tetrahydro-2-furoyl piperazine, 1-oxolan-2-ylcarbonyl piperazine, oxolan-2-yl piperazin-1-yl methanone | |
| oxolan-2-yl(piperazin-1-yl)methanone |
Specifications
| 63074-07-7 | |
| 97.5 | |
| 1.1700g/mL | |
| 5 g | |
| Glass bottle | |
| 1.5195 to 1.5215 | |
| 1.17 | |
| MFCD00190247 | |
| Solubility in water: miscible | |
| C1CC(OC1)C(=O)N2CCNCC2 | |
| 184.24 | |
| 184.24 | |
| 1-(Tetrahydro-2-furoyl)piperazine, 98% |
| Colorless to Yellow | |
| 100.0 | |
| 140°C (2.0 Mbar) | |
| Authentic | |
| Viscous Liquid | |
| 97.5% min. (GC) | |
| C9H16N2O2 | |
| 1-tetrahydro-2-furoyl piperazine, 1-2-tetrahydrofuroyl piperazine, piperazin-1-yl tetrahydrofuran-2-yl methanone, 1-2-tetrahydrofuroyl-piperazine, n-2-tetrahydrofuroyl piperazine, 1-oxolane-2-carbonyl piperazine, 1-tetrahydrofuran-2-ylcarbonyl piperazine, n-tetrahydro-2-furoyl piperazine, 1-oxolan-2-ylcarbonyl piperazine, oxolan-2-yl piperazin-1-yl methanone | |
| UKESBLFBQANJHH-UHFFFAOYSA-N | |
| oxolan-2-yl(piperazin-1-yl)methanone | |
| 2734641 | |
| 98% |
Safety and Handling
GHS H Statement
Causes severe skin burns and eye damage.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if pre
GHS Signal Word: Danger
RUO – Research Use Only