Learn More
1-Tetradecylphosphonic acid, 98%
CAS: 4671-75-4 | C14H31O3P | 278.373 g/mol
Supplier: Thermo Scientific Chemicals A1751106
Description
1-Tetradecylphosphonic acid is used to functionalize the surface of BaTiO3 nanoparticles.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications1-Tetradecylphosphonic acid is used to functionalize the surface of BaTiO3 nanoparticles.
Solubility
Soluble in ethanol, dimethyl sulfoxide and dimethyl formamide. Partially soluble in water.
Notes
Store in a cool place. Keep the container tightly closed in a dry and well-ventilated place. Incompatible withstrong oxidizing agents.
Specifications
| 1-Tetradecylphosphonic acid | |
| C14H31O3P | |
| 5 g | |
| Soluble in ethanol,dimethyl sulfoxide and dimethyl formamide. Partially soluble in water. | |
| CCCCCCCCCCCCCCP(=O)(O)O | |
| 278.373 | |
| 278.37 |
| 4671-75-4 | |
| MFCD00015834 | |
| phosphonic acid, tetradecyl, 1-tetradecanephosphonic acid, tetradecanephosphonic acid, tetradecyl phosphonic acid, n-tetradecylphosphonic acid, tetradecyl-phosphonic acid, 1-tetradecylphosphonic acid, phosphonic acid, p-tetradecyl, p-tetradecyl-phosphonic acid, tetradecylphosphonate | |
| BVQJQTMSTANITJ-UHFFFAOYSA-N | |
| tetradecylphosphonic acid | |
| 78401 | |
| 98% |
Chemical Identifiers
| 4671-75-4 | |
| 278.373 | |
| BVQJQTMSTANITJ-UHFFFAOYSA-N | |
| 78401 | |
| CCCCCCCCCCCCCCP(=O)(O)O |
| C14H31O3P | |
| MFCD00015834 | |
| phosphonic acid, tetradecyl, 1-tetradecanephosphonic acid, tetradecanephosphonic acid, tetradecyl phosphonic acid, n-tetradecylphosphonic acid, tetradecyl-phosphonic acid, 1-tetradecylphosphonic acid, phosphonic acid, p-tetradecyl, p-tetradecyl-phosphonic acid, tetradecylphosphonate | |
| tetradecylphosphonic acid |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 225-118-1
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only