missing translation for 'onlineSavingsMsg'
Learn More

(1-Tetradecyl)trimethylammonium bromide, 98%

Catalog No. AAL1029422
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1029422 100 g
AAL1029436 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1029422 Supplier Thermo Scientific Chemicals Supplier No. L1029422
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 1119-97-7 | C17H38BrN | 336.40 g/mol

Myristyltrimethylammonium has been used in a study to assess a surfactant-controlled synthetic method to obtain a nanophase of mesoporous ceria-zirconia solid solution containing cationic defects in the crystalline structure.  It has also been used in a study to determine the low molecular weight of organic acids in soils and plants by capillary zone electrophoresis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Myristyltrimethylammonium has been used in a study to assess a surfactant-controlled synthetic method to obtain a nanophase of mesoporous ceria-zirconia solid solution containing cationic defects in the crystalline structure.  It has also been used in a study to determine the low molecular weight of organic acids in soils and plants by capillary zone electrophoresis.

Solubility
Soluble in water. (100 g/L)

Notes
Light sensitive. Store in cool dry condition. Incompatible with strong oxidizing agents. Ensure ventilation.

Chemical Identifiers

CAS 1119-97-7
Molecular Formula C17H38BrN
Molecular Weight (g/mol) 336.40
MDL Number MFCD00011770
InChI Key CXRFDZFCGOPDTD-UHFFFAOYSA-M
Synonym cetrimide, tetradecyltrimethylammonium bromide, myristyltrimethylammonium bromide, tetradonium bromide, n,n,n-trimethyltetradecan-1-aminium bromide, tetradecyl trimethyl ammonium bromide, myrtrimonium bromide, mytab, mitmab, morpan t
PubChem CID 14250
ChEBI CHEBI:3565
IUPAC Name trimethyl(tetradecyl)azanium;bromide
SMILES [Br-].CCCCCCCCCCCCCC[N+](C)(C)C

Specifications

Melting Point 245°C to 250°C
Odor Odorless
Linear Formula CH3(CH2)13N(CH3)3Br
Quantity 100 g
UN Number UN1759
Beilstein 3633227
Merck Index 14,6336
Solubility Information Soluble in water. (100g/L)
Formula Weight 336.4
Percent Purity 98%
Chemical Name or Material (1-Tetradecyl)trimethylammonium bromide
Show More Show Less
Hazard Category H314-H335
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Hazard Class: 8; Packaging Group: II
EINECSNumber 214-291-9
RTECSNumber BS5776000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only