missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Tetradecanol 98.0+%, TCI America™
Supplier: TCI America T0084500ML
Specifications
| 1-Tetradecanol | |
| 38°C | |
| 173°C | |
| MFCD00004757 | |
| 3077 | |
| HLZKNKRTKFSKGZ-UHFFFAOYSA-N | |
| tetradecan-1-ol | |
| 8209 | |
| 214.39 | |
| Crystalline Lumps |
| 112-72-1 | |
| White | |
| C14H30O | |
| 500 mL | |
| 1-tetradecanol, myristyl alcohol, tetradecanol, tetradecyl alcohol, n-tetradecanol, myristic alcohol, n-tetradecyl alcohol, lanette k, loxanol v, lanette wax ks | |
| CCCCCCCCCCCCCCO | |
| 214.39 | |
| CHEBI:77417 | |
| ≥98.0% (GC) |
Chemical Identifiers
| 112-72-1 | |
| 214.39 | |
| HLZKNKRTKFSKGZ-UHFFFAOYSA-N | |
| 8209 | |
| tetradecan-1-ol |
| C14H30O | |
| MFCD00004757 | |
| 1-tetradecanol, myristyl alcohol, tetradecanol, tetradecyl alcohol, n-tetradecanol, myristic alcohol, n-tetradecyl alcohol, lanette k, loxanol v, lanette wax ks | |
| CHEBI:77417 | |
| CCCCCCCCCCCCCCO |
Safety and Handling
EINECSNumber : (2)-0217&(2)-3704
RTECSNumber : XB8655000
TSCA : Yes