missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Tetradecanol 98.0+%, TCI America™
$29.83 - $74.56
Chemical Identifiers
| CAS | 112-72-1 |
|---|---|
| Molecular Formula | C14H30O |
| Molecular Weight (g/mol) | 214.39 |
| MDL Number | MFCD00004757 |
| InChI Key | HLZKNKRTKFSKGZ-UHFFFAOYSA-N |
| Synonym | 1-tetradecanol, myristyl alcohol, tetradecanol, tetradecyl alcohol, n-tetradecanol, myristic alcohol, n-tetradecyl alcohol, lanette k, loxanol v, lanette wax ks |
| PubChem CID | 8209 |
| ChEBI | CHEBI:77417 |
| IUPAC Name | tetradecan-1-ol |
| SMILES | CCCCCCCCCCCCCCO |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
T008425ML
|
TCI America
T008425ML |
25 mL |
Each for $29.83
|
|
|||||
|
T0084500ML
|
TCI America
T0084500ML |
500 mL |
Each for $74.56
|
|
|||||
Chemical Identifiers
| 112-72-1 | |
| 214.39 | |
| HLZKNKRTKFSKGZ-UHFFFAOYSA-N | |
| 8209 | |
| tetradecan-1-ol |
| C14H30O | |
| MFCD00004757 | |
| 1-tetradecanol, myristyl alcohol, tetradecanol, tetradecyl alcohol, n-tetradecanol, myristic alcohol, n-tetradecyl alcohol, lanette k, loxanol v, lanette wax ks | |
| CHEBI:77417 | |
| CCCCCCCCCCCCCCO |
Specifications
| 112-72-1 | |
| White | |
| C14H30O | |
| 25 mL | |
| 1-tetradecanol, myristyl alcohol, tetradecanol, tetradecyl alcohol, n-tetradecanol, myristic alcohol, n-tetradecyl alcohol, lanette k, loxanol v, lanette wax ks | |
| CCCCCCCCCCCCCCO | |
| 214.39 | |
| CHEBI:77417 | |
| ≥98.0% (GC) | |
| 1-Tetradecanol |
| 38°C | |
| 173°C | |
| MFCD00004757 | |
| 3077 | |
| HLZKNKRTKFSKGZ-UHFFFAOYSA-N | |
| tetradecan-1-ol | |
| 8209 | |
| 214.39 | |
| Crystalline Lumps |
Safety and Handling
EINECSNumber : (2)-0217&(2)-3704
RTECSNumber : XB8655000
TSCA : Yes