Learn More
1-tert-Butylpiperazine, 97%
CAS: 38216-72-7 | C8H18N2 | 142.246 g/mol
$85.97 - $266.56
Chemical Identifiers
| CAS | 38216-72-7 |
|---|---|
| Molecular Formula | C8H18N2 |
| Molecular Weight (g/mol) | 142.246 |
| MDL Number | MFCD00049359 |
| InChI Key | PVMNSAIKFPWDQG-UHFFFAOYSA-N |
| Synonym | n-tert-butylpiperazine, 1-tert-butyl piperazine, n-t-butylpiperazine, 1-t-butyl piperazine, 1-tertbutyl piperazine, 1-tert-butyl-piperazine, piperazine, 1-1,1-dimethylethyl, n-tert-butyl piperazine, tert-butylpiperazine, tert-butyl piperazine |
| PubChem CID | 3530572 |
| IUPAC Name | 1-tert-butylpiperazine |
| SMILES | CC(C)(C)N1CCNCC1 |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAH6649703
|
Thermo Scientific Chemicals
H6649703 |
1 g |
Each for $85.97
|
|
|||||
|
AAH6649706
|
Thermo Scientific Chemicals
H6649706 |
5 g |
Each for $266.56
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 38216-72-7 | |
| 142.246 | |
| PVMNSAIKFPWDQG-UHFFFAOYSA-N | |
| 3530572 | |
| CC(C)(C)N1CCNCC1 |
| C8H18N2 | |
| MFCD00049359 | |
| n-tert-butylpiperazine, 1-tert-butyl piperazine, n-t-butylpiperazine, 1-t-butyl piperazine, 1-tertbutyl piperazine, 1-tert-butyl-piperazine, piperazine, 1-1,1-dimethylethyl, n-tert-butyl piperazine, tert-butylpiperazine, tert-butyl piperazine | |
| 1-tert-butylpiperazine |
Specifications
| 38216-72-7 | |
| Colorless to Yellow | |
| C8H18N2 | |
| 1 g | |
| Air, light, and moisture sensitive | |
| PVMNSAIKFPWDQG-UHFFFAOYSA-N | |
| 1-tert-butylpiperazine | |
| 3530572 | |
| 97% | |
| 1-tert-Butylpiperazine |
| 32°C to 35°C | |
| 85°C to 87°C (22 mmHg) | |
| MFCD00049359 | |
| UN3259 | |
| n-tert-butylpiperazine, 1-tert-butyl piperazine, n-t-butylpiperazine, 1-t-butyl piperazine, 1-tertbutyl piperazine, 1-tert-butyl-piperazine, piperazine, 1-1,1-dimethylethyl, n-tert-butyl piperazine, tert-butylpiperazine, tert-butyl piperazine | |
| CC(C)(C)N1CCNCC1 | |
| 142.246 | |
| 142.24 | |
| Low Melting Solid |
Safety and Handling
GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
H314-H335
DOTInformation : Transport Hazard Class: 8; Packaging Group: II; Proper Shipping Name: AMINES, SOLID, CORROSIVE, N.O.S.
TSCA : No
Recommended Storage : Ambient temperatures; Store under Argon
RUO – Research Use Only