accessibility menu, dialog, popup

UserName

CH3H3CCH3NNH

CAS RN 38216-72-7

IUPAC Name: 1-tert-butylpiperazine
Synonyms: 1-tert-butylpiperazine
Molecular Weight (g/mol): 142.25
Molecular Formula: C8H18N2
InChi Key: PVMNSAIKFPWDQG-UHFFFAOYSA-N
SMILES: CC(C)(C)N1CCNCC1
Boiling Point 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)