missing translation for 'onlineSavingsMsg'
Learn More

1-tert-Butylpiperazine, 97%

Catalog No. AAH6649706
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6649706 5 g
AAH6649703 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6649706 Supplier Thermo Scientific Chemicals Supplier No. H6649706
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 38216-72-7 | C8H18N2 | 142.246 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 38216-72-7
Molecular Formula C8H18N2
Molecular Weight (g/mol) 142.246
MDL Number MFCD00049359
InChI Key PVMNSAIKFPWDQG-UHFFFAOYSA-N
Synonym n-tert-butylpiperazine, 1-tert-butyl piperazine, n-t-butylpiperazine, 1-t-butyl piperazine, 1-tertbutyl piperazine, 1-tert-butyl-piperazine, piperazine, 1-1,1-dimethylethyl, n-tert-butyl piperazine, tert-butylpiperazine, tert-butyl piperazine
PubChem CID 3530572
IUPAC Name 1-tert-butylpiperazine
SMILES CC(C)(C)N1CCNCC1

Specifications

Melting Point 32°C to 35°C
Color Colorless to Yellow
Boiling Point 85°C to 87°C (22 mmHg)
Quantity 5 g
UN Number UN3259
Sensitivity Air, light, and moisture sensitive
Formula Weight 142.24
Percent Purity 97%
Physical Form Low Melting Solid
Chemical Name or Material 1-tert-Butylpiperazine
Hazard Category H314-H335
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packaging Group: II; Proper Shipping Name: AMINES, SOLID, CORROSIVE, N.O.S.
TSCA No
Recommended Storage Ambient temperatures; Store under Argon

RUO – Research Use Only