missing translation for 'onlineSavingsMsg'
Learn More

1-(tert-Butyldimethylsilyl)imidazole, 97%

CAS: 54925-64-3 | C9H18N2Si | 182.34 g/mol

Supplier:  Thermo Scientific Chemicals 04146206

 View more versions of this product

Catalog No. AA4146206


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

1-(tert-Butyldimethylsilyl)imidazole
0.94 g/mL
99°C (210°F)
1.4805
5 g
1-tert-butyldimethylsilyl-1h-imidazole, 1-tert-butyldimethylsilyl imidazole, tert-butyldimethylsilylimidazole, n-tert-butyldimethylsilylimidazole, t-butyldimethylsilylimidazole, 1-t-butyldimethylsilyl imidazole, 1h-imidazole, 1-1,1-dimethylethyl dimethylsilyl, tbdmsim, imidazole, tbdms derivative, 1-tert-butyldimethylsilylimidazole
CC(C)(C)[Si](C)(C)N1C=CN=C1
182.34
182.34
Liquid
54925-64-3
247°C to 250°C
C9H18N2Si
MFCD00011682
Moisture sensitive
VUENSYJCBOSTCS-UHFFFAOYSA-N
tert-butyl-imidazol-1-yl-dimethylsilane
171385
97%

Chemical Identifiers

54925-64-3
182.34
VUENSYJCBOSTCS-UHFFFAOYSA-N
171385
CC(C)(C)[Si](C)(C)N1C=CN=C1
C9H18N2Si
MFCD00011682
1-tert-butyldimethylsilyl-1h-imidazole, 1-tert-butyldimethylsilyl imidazole, tert-butyldimethylsilylimidazole, n-tert-butyldimethylsilylimidazole, t-butyldimethylsilylimidazole, 1-t-butyldimethylsilyl imidazole, 1h-imidazole, 1-1,1-dimethylethyl dimethylsilyl, tbdmsim, imidazole, tbdms derivative, 1-tert-butyldimethylsilylimidazole
tert-butyl-imidazol-1-yl-dimethylsilane
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 259-398-1

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only