missing translation for 'onlineSavingsMsg'
Learn More

1-(tert-Butyldimethylsilyl)imidazole, 97%

Catalog No. p-7051535
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
1 g
25 g
5 g
Catalog No. Quantity
AA4146203 1 g
AA4146206 5 g
AA4146214 25 g
3 options

Catalog No. AA4146203

Supplier: Thermo Scientific Chemicals 04146203

Only null left

CAS: 54925-64-3 | C9H18N2Si | 182.34 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 54925-64-3
Molecular Formula C9H18N2Si
Molecular Weight (g/mol) 182.34
MDL Number MFCD00011682
InChI Key VUENSYJCBOSTCS-UHFFFAOYSA-N
Synonym 1-tert-butyldimethylsilyl-1h-imidazole, 1-tert-butyldimethylsilyl imidazole, tert-butyldimethylsilylimidazole, n-tert-butyldimethylsilylimidazole, t-butyldimethylsilylimidazole, 1-t-butyldimethylsilyl imidazole, 1h-imidazole, 1-1,1-dimethylethyl dimethylsilyl, tbdmsim, imidazole, tbdms derivative, 1-tert-butyldimethylsilylimidazole
PubChem CID 171385
IUPAC Name tert-butyl-imidazol-1-yl-dimethylsilane
SMILES CC(C)(C)[Si](C)(C)N1C=CN=C1

Specifications

Density 0.94 g/mL
Boiling Point 247°C to 250°C
Flash Point 99°C (210°F)
Refractive Index 1.4805
Quantity 1 g
Sensitivity Moisture sensitive
Formula Weight 182.34
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 1-(tert-Butyldimethylsilyl)imidazole
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 259-398-1
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only