missing translation for 'onlineSavingsMsg'
Learn More

1-tert-Butyl N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-L-aspartate 98.0+%, TCI America™

Catalog No. p-7129715
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F05031G 1 g
F05035G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F05031G Supplier TCI America Supplier No. F05031G
Only null left

Chemical Identifiers

CAS 129460-09-9
Molecular Formula C23H25NO6
Molecular Weight (g/mol) 411.45
MDL Number MFCD00065631
InChI Key VZXQYACYLGRQJU-UHFFFAOYNA-N
Synonym fmoc-asp-otbu, fmoc-asp-obut, s-3-9h-fluoren-9-yl methoxy carbonyl amino-4-tert-butoxy-4-oxobutanoic acid, l-fmoc-aspartic acid alpha-t-butyl ester, fmoc-asp oh-otbu, n-fmoc-l-aspartic acid 1-tert-butyl ester, 1-tert-butyl n-9h-fluoren-9-ylmethoxy carbonyl-l-aspartate, 3s-4-tert-butoxy-3-9h-fluoren-9-ylmethoxy carbonyl amino-4-oxobutanoic acid, 1-tert-butyl n-fmoc-l-aspartate, fmoc asp-otbu
PubChem CID 7017910
IUPAC Name 4-(tert-butoxy)-3-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)-4-oxobutanoic acid
SMILES CC(C)(C)OC(=O)C(CC(O)=O)NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12

Specifications

Color White-Yellow
Quantity 1 g
Formula Weight 411.45
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 1-tert-Butyl N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-L-aspartate