missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-tert-Butyl N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-L-aspartate 98.0+%, TCI America™
Specifications
Specifications
| Color | White-Yellow |
| Quantity | 1 g |
| Formula Weight | 411.45 |
| Percent Purity | ≥98.0% (HPLC,T) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | 1-tert-Butyl N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-L-aspartate |
Safety and Handling
| TSCA | No |