missing translation for 'onlineSavingsMsg'
Learn More

1-tert-Butyl 3-Ethyl 4-Oxopyrrolidine-1,3-dicarboxylate 97.0+%, TCI America™

Catalog No. B46585G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B46585G 5 g
B46581G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B46585G Supplier TCI America Supplier No. B46585G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 146256-98-6
Molecular Formula C12H19NO5
Molecular Weight (g/mol) 257.286
MDL Number MFCD06656201
InChI Key CWRFSVAKVCSOPI-UHFFFAOYSA-N
Synonym 4-Oxopyrrolidine-1,3-dicarboxylic Acid 1-tert-Butyl 3-Ethyl Ester
PubChem CID 10868942
IUPAC Name 1-O-tert-butyl 3-O-ethyl 4-oxopyrrolidine-1,3-dicarboxylate
SMILES CCOC(=O)C1CN(CC1=O)C(=O)OC(C)(C)C

Specifications

Melting Point 55°C
Color Yellow
Boiling Point 120°C
Quantity 5 g
Formula Weight 257.29
Percent Purity ≥97.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material 1-tert-Butyl 3-Ethyl 4-Oxopyrrolidine-1,3-dicarboxylate
TSCA No
Recommended Storage Refrigerator