accessibility menu, dialog, popup

UserName

H3COONOOOCH3H3CCH3

CAS RN 146256-98-6

IUPAC Name: 1-tert-butyl 3-ethyl 4-oxopyrrolidine-1,3-dicarboxylate
Synonyms: 1-tert-butyl 3-ethyl 4-oxopyrrolidine-1,3-dicarboxylate
Molecular Weight (g/mol): 257.29
Molecular Formula: C12H19NO5
InChi Key: CWRFSVAKVCSOPI-UHFFFAOYNA-N
SMILES: CCOC(=O)C1CN(CC1=O)C(=O)OC(C)(C)C
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results