missing translation for 'onlineSavingsMsg'
Learn More
Learn More
[1-(tert-Butoxycarbonylamino)cyclopropyl]methanol 97.0+%, TCI America™
$59.69 - $190.59
Chemical Identifiers
| CAS | 107017-73-2 |
|---|---|
| Molecular Formula | C9H17NO3 |
| Molecular Weight (g/mol) | 187.239 |
| MDL Number | MFCD09749954 |
| InChI Key | HFMAZNJKNNRONT-UHFFFAOYSA-N |
| Synonym | [1-(Boc-amino)cyclopropyl]methanol, tert-Butyl [1-(Hydroxymethyl)cyclopropyl]carbamate, [1-(Hydroxymethyl)cyclopropyl]carbamic Acid tert-Butyl Ester |
| PubChem CID | 11286907 |
| IUPAC Name | tert-butyl N-[1-(hydroxymethyl)cyclopropyl]carbamate |
| SMILES | CC(C)(C)OC(=O)NC1(CC1)CO |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
B4251200MG
|
TCI America
B4251200MG |
200 mg |
Each for $59.69
|
|
|||||
|
B42511G
|
TCI America
B42511G |
1 g |
Each for $190.59
|
|
|||||
Chemical Identifiers
| 107017-73-2 | |
| 187.239 | |
| HFMAZNJKNNRONT-UHFFFAOYSA-N | |
| 11286907 | |
| CC(C)(C)OC(=O)NC1(CC1)CO |
| C9H17NO3 | |
| MFCD09749954 | |
| [1-(Boc-amino)cyclopropyl]methanol, tert-Butyl [1-(Hydroxymethyl)cyclopropyl]carbamate, [1-(Hydroxymethyl)cyclopropyl]carbamic Acid tert-Butyl Ester | |
| tert-butyl N-[1-(hydroxymethyl)cyclopropyl]carbamate |
Specifications
| 83°C | |
| Yellow | |
| MFCD09749954 | |
| HFMAZNJKNNRONT-UHFFFAOYSA-N | |
| tert-butyl N-[1-(hydroxymethyl)cyclopropyl]carbamate | |
| 187.239 | |
| 187.24 | |
| Crystalline Powder |
| 107017-73-2 | |
| C9H17NO3 | |
| [1-(Boc-amino)cyclopropyl]methanol, tert-Butyl [1-(Hydroxymethyl)cyclopropyl]carbamate, [1-(Hydroxymethyl)cyclopropyl]carbamic Acid tert-Butyl Ester | |
| CC(C)(C)OC(=O)NC1(CC1)CO | |
| 200 mg | |
| 11286907 | |
| ≥97.0% (GC) | |
| [1-(tert-Butoxycarbonylamino)cyclopropyl]methanol |
Safety and Handling
TSCA : No