missing translation for 'onlineSavingsMsg'
Learn More

[1-(tert-Butoxycarbonylamino)cyclopropyl]methanol 97.0+%, TCI America™

Catalog No. b4251200mg
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B4251200MG 200 mg
B42511G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B4251200MG Supplier TCI America Supplier No. B4251200MG
Only null left

Chemical Identifiers

CAS 107017-73-2
Molecular Formula C9H17NO3
Molecular Weight (g/mol) 187.239
MDL Number MFCD09749954
InChI Key HFMAZNJKNNRONT-UHFFFAOYSA-N
Synonym [1-(Boc-amino)cyclopropyl]methanol, tert-Butyl [1-(Hydroxymethyl)cyclopropyl]carbamate, [1-(Hydroxymethyl)cyclopropyl]carbamic Acid tert-Butyl Ester
PubChem CID 11286907
IUPAC Name tert-butyl N-[1-(hydroxymethyl)cyclopropyl]carbamate
SMILES CC(C)(C)OC(=O)NC1(CC1)CO

Specifications

Melting Point 83°C
Color Yellow
Quantity 200 mg
Formula Weight 187.24
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material [1-(tert-Butoxycarbonylamino)cyclopropyl]methanol