missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-(tert-Butoxycarbonyl)guanidine 98.0+%, TCI America™
Supplier: TCI America B500125G
Specifications
| 1-(tert-Butoxycarbonyl)guanidine | |
| 168°C | |
| C6H13N3O2 | |
| 25 g | |
| UMOZLQVSOVNSCA-UHFFFAOYSA-N | |
| tert-butyl N-(diaminomethylidene)carbamate | |
| 11378649 | |
| ≥98.0% (T) |
| 219511-71-4 | |
| White | |
| MFCD07369706 | |
| boc-guanidine, n-boc-guanidine, 1-tert-butoxycarbonyl guanidine, tert-butyl n-carbamimidoylcarbamate, boc guanidine, 1-boc-guanidine, tert-butyl n-diaminomethylidene carbamate, tert-butoxycarbonylguanidine, acmc-209x9z | |
| CC(C)(C)OC(=O)N=C(N)N | |
| 159.189 | |
| 159.19 | |
| Crystalline Powder |
Chemical Identifiers
| 219511-71-4 | |
| 159.189 | |
| UMOZLQVSOVNSCA-UHFFFAOYSA-N | |
| 11378649 | |
| CC(C)(C)OC(=O)N=C(N)N |
| C6H13N3O2 | |
| MFCD07369706 | |
| boc-guanidine, n-boc-guanidine, 1-tert-butoxycarbonyl guanidine, tert-butyl n-carbamimidoylcarbamate, boc guanidine, 1-boc-guanidine, tert-butyl n-diaminomethylidene carbamate, tert-butoxycarbonylguanidine, acmc-209x9z | |
| tert-butyl N-(diaminomethylidene)carbamate |
Safety and Handling
TSCA : No