missing translation for 'onlineSavingsMsg'
Learn More

1-(tert-Butoxycarbonyl)guanidine 98.0+%, TCI America™

Catalog No. p-7123892
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B50015G 5 g
B500125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B50015G Supplier TCI America Supplier No. B50015G
Only null left

Chemical Identifiers

CAS 219511-71-4
Molecular Formula C6H13N3O2
Molecular Weight (g/mol) 159.189
MDL Number MFCD07369706
InChI Key UMOZLQVSOVNSCA-UHFFFAOYSA-N
Synonym boc-guanidine, n-boc-guanidine, 1-tert-butoxycarbonyl guanidine, tert-butyl n-carbamimidoylcarbamate, boc guanidine, 1-boc-guanidine, tert-butyl n-diaminomethylidene carbamate, tert-butoxycarbonylguanidine, acmc-209x9z
PubChem CID 11378649
IUPAC Name tert-butyl N-(diaminomethylidene)carbamate
SMILES CC(C)(C)OC(=O)N=C(N)N

Specifications

Melting Point 168°C
Color White
Quantity 5 g
Formula Weight 159.19
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 1-(tert-Butoxycarbonyl)guanidine