missing translation for 'onlineSavingsMsg'
Learn More

1-(tert-Butoxycarbonyl)azetidine-3-carboxylic Acid 98.0+%, TCI America™

Catalog No. B35405G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B35405G 5 g
B35401G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B35405G Supplier TCI America Supplier No. B35405G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 142253-55-2
Molecular Formula C9H14NO4
Molecular Weight (g/mol) 200.22
MDL Number MFCD01860897
InChI Key NCADHSLPNSTDMJ-UHFFFAOYSA-M
Synonym 1-boc-azetidine-3-carboxylic acid, 1-n-boc-3-azetidinecarboxylic acid, n-boc-azetidine-3-carboxylic acid, boc-azetidine-3-carboxylic acid, 1-tert-butoxycarbonyl azetidine-3-carboxylic acid, 1-tert-butoxy carbonyl azetidine-3-carboxylic acid, 1-n-boc-azetidine-3-carboxylic acid, boc-aze 3-oh, azetidine-1,3-dicarboxylic acid mono-tert-butyl ester, n-boc-3-azetidine carboxylic acid
PubChem CID 2755981
IUPAC Name 1-[(tert-butoxy)carbonyl]azetidine-3-carboxylate
SMILES CC(C)(C)OC(=O)N1CC(C1)C([O-])=O

Specifications

Melting Point 105°C
Color White-Yellow
Quantity 5 g
Formula Weight 201.22
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 1-(tert-Butoxycarbonyl)azetidine-3-carboxylic Acid