accessibility menu, dialog, popup

UserName

CH3H3CCH3OONOO

CAS RN 142253-55-2

IUPAC Name: 1-[(tert-butoxy)carbonyl]azetidine-3-carboxylate
Synonyms: 1-(tert-butoxycarbonyl)azetidine-3-carboxylate
Molecular Weight (g/mol): 200.22
Molecular Formula: C9H14NO4
InChi Key: NCADHSLPNSTDMJ-UHFFFAOYSA-M
SMILES: CC(C)(C)OC(=O)N1CC(C1)C([O-])=O
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)