missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-(tert-Butoxycarbonyl)azetidine-3-carboxaldehyde 98.0+%, TCI America™
$79.74 - $239.23
Chemical Identifiers
| CAS | 177947-96-5 |
|---|---|
| Molecular Formula | C9H15NO3 |
| Molecular Weight (g/mol) | 185.223 |
| MDL Number | MFCD06656142 |
| InChI Key | JVQOZRRUGOADSU-UHFFFAOYSA-N |
| Synonym | 1-boc-3-azetidinecarboxaldehyde, 1-boc-3-formylazetidine, 1-boc-azetidine-3-carboxaldehyde, azetidine-3-carboxaldehyde, 1-azetidinecarboxylic acid, 3-formyl-, 1,1-dimethylethyl ester, 1-tert-butoxycarbonyl azetidine-3-carboxaldehyde, 3-formylazetidine-1-carboxylic acid tert-butylester, pubchem10136, 1-boc-3-azetidinecarbaldehyde, 1-boc-3-azetidine carboxaldehyde |
| PubChem CID | 10726182 |
| IUPAC Name | tert-butyl 3-formylazetidine-1-carboxylate |
| SMILES | CC(C)(C)OC(=O)N1CC(C1)C=O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
B5160200MG
|
TCI America
B5160200MG |
200 mg |
Each for $79.74
|
|
|||||
|
B51601G
|
TCI America
B51601G |
1 g |
Each for $239.23
|
|
|||||
Chemical Identifiers
| 177947-96-5 | |
| 185.223 | |
| JVQOZRRUGOADSU-UHFFFAOYSA-N | |
| 10726182 | |
| CC(C)(C)OC(=O)N1CC(C1)C=O |
| C9H15NO3 | |
| MFCD06656142 | |
| 1-boc-3-azetidinecarboxaldehyde, 1-boc-3-formylazetidine, 1-boc-azetidine-3-carboxaldehyde, azetidine-3-carboxaldehyde, 1-azetidinecarboxylic acid, 3-formyl-, 1,1-dimethylethyl ester, 1-tert-butoxycarbonyl azetidine-3-carboxaldehyde, 3-formylazetidine-1-carboxylic acid tert-butylester, pubchem10136, 1-boc-3-azetidinecarbaldehyde, 1-boc-3-azetidine carboxaldehyde | |
| tert-butyl 3-formylazetidine-1-carboxylate |
Specifications
| 177947-96-5 | |
| Yellow | |
| MFCD06656142 | |
| JVQOZRRUGOADSU-UHFFFAOYSA-N | |
| tert-butyl 3-formylazetidine-1-carboxylate | |
| 185.223 | |
| 185.22 | |
| Liquid |
| 82°C | |
| C9H15NO3 | |
| 1-boc-3-azetidinecarboxaldehyde, 1-boc-3-formylazetidine, 1-boc-azetidine-3-carboxaldehyde, azetidine-3-carboxaldehyde, 1-azetidinecarboxylic acid, 3-formyl-, 1,1-dimethylethyl ester, 1-tert-butoxycarbonyl azetidine-3-carboxaldehyde, 3-formylazetidine-1-carboxylic acid tert-butylester, pubchem10136, 1-boc-3-azetidinecarbaldehyde, 1-boc-3-azetidine carboxaldehyde | |
| CC(C)(C)OC(=O)N1CC(C1)C=O | |
| 200 mg | |
| 10726182 | |
| ≥98.0% (GC) | |
| 1-(tert-Butoxycarbonyl)azetidine-3-carboxaldehyde |
Safety and Handling
TSCA : No