missing translation for 'onlineSavingsMsg'
Learn More

1-(tert-Butoxycarbonyl)azetidine-3-carboxaldehyde 98.0+%, TCI America™

Catalog No. B5160200MG
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B5160200MG 200 mg
B51601G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B5160200MG Supplier TCI America Supplier No. B5160200MG

Chemical Identifiers

CAS 177947-96-5
Molecular Formula C9H15NO3
Molecular Weight (g/mol) 185.223
MDL Number MFCD06656142
InChI Key JVQOZRRUGOADSU-UHFFFAOYSA-N
Synonym 1-boc-3-azetidinecarboxaldehyde, 1-boc-3-formylazetidine, 1-boc-azetidine-3-carboxaldehyde, azetidine-3-carboxaldehyde, 1-azetidinecarboxylic acid, 3-formyl-, 1,1-dimethylethyl ester, 1-tert-butoxycarbonyl azetidine-3-carboxaldehyde, 3-formylazetidine-1-carboxylic acid tert-butylester, pubchem10136, 1-boc-3-azetidinecarbaldehyde, 1-boc-3-azetidine carboxaldehyde
PubChem CID 10726182
IUPAC Name tert-butyl 3-formylazetidine-1-carboxylate
SMILES CC(C)(C)OC(=O)N1CC(C1)C=O

Specifications

Boiling Point 82°C
Color Yellow
Quantity 200 mg
Formula Weight 185.22
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 1-(tert-Butoxycarbonyl)azetidine-3-carboxaldehyde