missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-(tert-Butoxycarbonyl)-4-piperidinemethanol 98.0+%, TCI America™
$31.68 - $180.81
Chemical Identifiers
| CAS | 123855-51-6 |
|---|---|
| Molecular Formula | C11H21NO3 |
| Molecular Weight (g/mol) | 215.29 |
| MDL Number | MFCD02094488 |
| InChI Key | CTEDVGRUGMPBHE-UHFFFAOYSA-N |
| Synonym | n-boc-4-piperidinemethanol, tert-butyl 4-hydroxymethyl piperidine-1-carboxylate, 1-boc-4-piperidinemethanol, n-boc-4-hydroxymethylpiperidine, tert-butyl 4-hydroxymethyl-1-piperidinecarboxylate, 1-tert-butoxycarbonyl-4-hydroxymethyl piperidine, 1-n-boc-4-hydroxymethyl-piperidine, n-boc-4-hydroxylmethylene-piperidine, 1-tert-butoxycarbonyl-4-piperidinemethanol, n-tert-butoxycarbonyl-4-piperidinemethanol |
| PubChem CID | 2764081 |
| IUPAC Name | tert-butyl 4-(hydroxymethyl)piperidine-1-carboxylate |
| SMILES | CC(C)(C)OC(=O)N1CCC(CO)CC1 |
Chemical Identifiers
| 123855-51-6 | |
| 215.29 | |
| CTEDVGRUGMPBHE-UHFFFAOYSA-N | |
| 2764081 | |
| CC(C)(C)OC(=O)N1CCC(CO)CC1 |
| C11H21NO3 | |
| MFCD02094488 | |
| n-boc-4-piperidinemethanol, tert-butyl 4-hydroxymethyl piperidine-1-carboxylate, 1-boc-4-piperidinemethanol, n-boc-4-hydroxymethylpiperidine, tert-butyl 4-hydroxymethyl-1-piperidinecarboxylate, 1-tert-butoxycarbonyl-4-hydroxymethyl piperidine, 1-n-boc-4-hydroxymethyl-piperidine, n-boc-4-hydroxylmethylene-piperidine, 1-tert-butoxycarbonyl-4-piperidinemethanol, n-tert-butoxycarbonyl-4-piperidinemethanol | |
| tert-butyl 4-(hydroxymethyl)piperidine-1-carboxylate |
Specifications
| 123855-51-6 | |
| White-Yellow | |
| MFCD02094488 | |
| n-boc-4-piperidinemethanol, tert-butyl 4-hydroxymethyl piperidine-1-carboxylate, 1-boc-4-piperidinemethanol, n-boc-4-hydroxymethylpiperidine, tert-butyl 4-hydroxymethyl-1-piperidinecarboxylate, 1-tert-butoxycarbonyl-4-hydroxymethyl piperidine, 1-n-boc-4-hydroxymethyl-piperidine, n-boc-4-hydroxylmethylene-piperidine, 1-tert-butoxycarbonyl-4-piperidinemethanol, n-tert-butoxycarbonyl-4-piperidinemethanol | |
| CC(C)(C)OC(=O)N1CCC(CO)CC1 | |
| 215.29 | |
| 215.29 | |
| Crystalline Powder |
| 79°C | |
| C11H21NO3 | |
| 1 g | |
| CTEDVGRUGMPBHE-UHFFFAOYSA-N | |
| tert-butyl 4-(hydroxymethyl)piperidine-1-carboxylate | |
| 2764081 | |
| ≥98.0% (GC) | |
| 1-(tert-Butoxycarbonyl)-4-piperidinemethanol |
Safety and Handling
TSCA : No