missing translation for 'onlineSavingsMsg'
Learn More

1-(tert-Butoxycarbonyl)-4-piperidinecarboxaldehyde 95.0+%, TCI America™

Catalog No. b38731g
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
1 g
5 g
Catalog No. Quantity
B38731G 1 g
B38735G 5 g
2 options

Catalog No. B38731G

Supplier: TCI America B38731G

Only null left

Chemical Identifiers

CAS 137076-22-3
Molecular Formula C11H19NO3
Molecular Weight (g/mol) 213.28
MDL Number MFCD02179019
InChI Key JYUQEWCJWDGCRX-UHFFFAOYSA-N
Synonym 1-boc-4-piperidinecarboxaldehyde, 1-boc-4-formylpiperidine, 1-tert-butoxycarbonyl-4-piperidinecarboxaldehyde, 1-boc-piperidine-4-carboxaldehyde, n-boc-4-formylpiperidine, n-boc-4-piperidinecarboxaldehyde, 1-tert-butoxycarbonyl-4-formylpiperidine, 4-formyl-piperidine-1-carboxylic acid tert-butyl ester, n-boc-piperidine-4-carbaldehyde, 4-formylpiperidine-1-carboxylic acid tert-butyl ester
PubChem CID 1514430
IUPAC Name tert-butyl 4-formylpiperidine-1-carboxylate
SMILES CC(C)(C)OC(=O)N1CCC(CC1)C=O

Specifications

Melting Point 35°C
Color White-Yellow
Quantity 1 g
Formula Weight 213.28
Percent Purity ≥95.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1-(tert-Butoxycarbonyl)-4-piperidinecarboxaldehyde
TSCA No
Recommended Storage Refrigerator