accessibility menu, dialog, popup

UserName

CH3H3CCH3OONO

CAS RN 137076-22-3

IUPAC Name: tert-butyl 4-formylpiperidine-1-carboxylate
Synonyms: tert-butyl 4-formylpiperidine-1-carboxylate
Molecular Weight (g/mol): 213.28
Molecular Formula: C11H19NO3
InChi Key: JYUQEWCJWDGCRX-UHFFFAOYSA-N
SMILES: CC(C)(C)OC(=O)N1CCC(CC1)C=O
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)