missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-(tert-Butoxycarbonyl)-4-(p-toluenesulfonyloxy)piperidine 98.0+%, TCI America™
$70.84 - $70.84
Chemical Identifiers
| CAS | 118811-07-7 |
|---|---|
| Molecular Formula | C17H25NO5S |
| Molecular Weight (g/mol) | 355.449 |
| MDL Number | MFCD06796535 |
| InChI Key | IKOMRHLHPZAEMV-UHFFFAOYSA-N |
| Synonym | 1-Boc-4-tosyloxypiperidine, tert-Butyl 4-(p-Toluenesulfonyloxy)piperidine-1-carboxylate, 4-(p-Toluenesulfonyloxy)piperidine-1-carboxylic Acid tert-Butyl Ester, tert-Butyl 4-Tosyloxypiperidine-1-carboxylate, 4-Tosyloxypiperidine-1-carboxylic Acid tert-Buty |
| PubChem CID | 15734713 |
| IUPAC Name | tert-butyl 4-(4-methylphenyl)sulfonyloxypiperidine-1-carboxylate |
| SMILES | CC1=CC=C(C=C1)S(=O)(=O)OC2CCN(CC2)C(=O)OC(C)(C)C |
Chemical Identifiers
| 118811-07-7 | |
| 355.449 | |
| IKOMRHLHPZAEMV-UHFFFAOYSA-N | |
| 15734713 | |
| CC1=CC=C(C=C1)S(=O)(=O)OC2CCN(CC2)C(=O)OC(C)(C)C |
| C17H25NO5S | |
| MFCD06796535 | |
| 1-Boc-4-tosyloxypiperidine, tert-Butyl 4-(p-Toluenesulfonyloxy)piperidine-1-carboxylate, 4-(p-Toluenesulfonyloxy)piperidine-1-carboxylic Acid tert-Butyl Ester, tert-Butyl 4-Tosyloxypiperidine-1-carboxylate, 4-Tosyloxypiperidine-1-carboxylic Acid tert-Buty | |
| tert-butyl 4-(4-methylphenyl)sulfonyloxypiperidine-1-carboxylate |
Specifications
| 118811-07-7 | |
| White | |
| MFCD06796535 | |
| 1-Boc-4-tosyloxypiperidine, tert-Butyl 4-(p-Toluenesulfonyloxy)piperidine-1-carboxylate, 4-(p-Toluenesulfonyloxy)piperidine-1-carboxylic Acid tert-Butyl Ester, tert-Butyl 4-Tosyloxypiperidine-1-carboxylate, 4-Tosyloxypiperidine-1-carboxylic Acid tert-Buty | |
| CC1=CC=C(C=C1)S(=O)(=O)OC2CCN(CC2)C(=O)OC(C)(C)C | |
| 355.449 | |
| 355.45 | |
| Crystalline Powder |
| 98°C | |
| C17H25NO5S | |
| 1 g | |
| IKOMRHLHPZAEMV-UHFFFAOYSA-N | |
| tert-butyl 4-(4-methylphenyl)sulfonyloxypiperidine-1-carboxylate | |
| 15734713 | |
| ≥98.0% (HPLC) | |
| 1-(tert-Butoxycarbonyl)-4-(p-toluenesulfonyloxy)piperidine |
Safety and Handling
TSCA : No