CAS RN 118811-07-7
IUPAC Name:
tert-butyl 4-[(4-methylbenzenesulfonyl)oxy]piperidine-1-carboxylate
Synonyms:
tert-butyl 4-[(4-methylbenzenesulfonyl)oxy]piperidine-1-carboxylate
Molecular Weight (g/mol):
355.45
Molecular Formula:
C17H25NO5S
InChi Key:
IKOMRHLHPZAEMV-UHFFFAOYSA-N
SMILES:
CC1=CC=C(C=C1)S(=O)(=O)OC1CCN(CC1)C(=O)OC(C)(C)C
1
–
1
of
1
results
1
1-(tert-Butoxycarbonyl)-4-(p-toluenesulfonyloxy)piperidine 98.0+%, TCI America™
CAS: 118811-07-7 Molecular Formula: C17H25NO5S Molecular Weight (g/mol): 355.449 MDL Number: MFCD06796535 InChI Key: IKOMRHLHPZAEMV-UHFFFAOYSA-N Synonym: 1-Boc-4-tosyloxypiperidine, tert-Butyl 4-(p-Toluenesulfonyloxy)piperidine-1-carboxylate, 4-(p-Toluenesulfonyloxy)piperidine-1-carboxylic Acid tert-Butyl Ester, tert-Butyl 4-Tosyloxypiperidine-1-carboxylate, 4-Tosyloxypiperidine-1-carboxylic Acid tert-Buty PubChem CID: 15734713 IUPAC Name: tert-butyl 4-(4-methylphenyl)sulfonyloxypiperidine-1-carboxylate SMILES: CC1=CC=C(C=C1)S(=O)(=O)OC2CCN(CC2)C(=O)OC(C)(C)C