missing translation for 'onlineSavingsMsg'
Learn More

1-(tert-Butoxycarbonyl)-4-(p-toluenesulfonyloxy)piperidine 98.0+%, TCI America™

Catalog No. B45161G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B45161G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B45161G Supplier TCI America Supplier No. B45161G
Only null left

Chemical Identifiers

CAS 118811-07-7
Molecular Formula C17H25NO5S
Molecular Weight (g/mol) 355.449
MDL Number MFCD06796535
InChI Key IKOMRHLHPZAEMV-UHFFFAOYSA-N
Synonym 1-Boc-4-tosyloxypiperidine, tert-Butyl 4-(p-Toluenesulfonyloxy)piperidine-1-carboxylate, 4-(p-Toluenesulfonyloxy)piperidine-1-carboxylic Acid tert-Butyl Ester, tert-Butyl 4-Tosyloxypiperidine-1-carboxylate, 4-Tosyloxypiperidine-1-carboxylic Acid tert-Buty
PubChem CID 15734713
IUPAC Name tert-butyl 4-(4-methylphenyl)sulfonyloxypiperidine-1-carboxylate
SMILES CC1=CC=C(C=C1)S(=O)(=O)OC2CCN(CC2)C(=O)OC(C)(C)C

Specifications

Melting Point 98°C
Color White
Quantity 1 g
Formula Weight 355.45
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 1-(tert-Butoxycarbonyl)-4-(p-toluenesulfonyloxy)piperidine