missing translation for 'onlineSavingsMsg'
Learn More

1-(tert-Butoxycarbonyl)-4-[(p-toluenesulfonyloxy)methyl]piperidine 98.0+%, TCI America™

Catalog No. p-7123730
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B47595G 5 g
B475925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B47595G Supplier TCI America Supplier No. B47595G

Chemical Identifiers

CAS 166815-96-9
Molecular Formula C18H27NO5S
Molecular Weight (g/mol) 369.476
MDL Number MFCD05864740
InChI Key DARTVAOOTJKHQW-UHFFFAOYSA-N
Synonym n-boc-4-4-toluenesulfonyloxymethyl piperidine, tert-butyl 4-tosyloxy methyl piperidine-1-carboxylate, n-tert-butoxycarbonyl-4-4-toluenesulfonyloxymethyl piperidine, 1-boc-4-tosyloxymethyl piperidine, tert-butyl 4-4-methylbenzenesulfonyl oxy methyl piperidine-1-carboxylate, 4-toluene-4-sulfonyloxymethyl-piperidine-1-carboxylic acid tert-butyl ester, tert-butyl 4-4-methylphenyl sulfonyl oxy methyl piperidine-1-carboxylate, 1-tert-butoxycarbonyl-4-p-toluenesulfonyloxy methyl piperidine, 1-tert-butoxycarbonyl-4-tosyloxymethyl piperidine, tert-butyl 4-tosyloxymethyl piperidine-1-carboxylate
PubChem CID 10642851
IUPAC Name tert-butyl 4-[(4-methylphenyl)sulfonyloxymethyl]piperidine-1-carboxylate
SMILES CC1=CC=C(C=C1)S(=O)(=O)OCC2CCN(CC2)C(=O)OC(C)(C)C

Specifications

Melting Point 74°C
Color White-Yellow
Quantity 5 g
Formula Weight 369.48
Percent Purity ≥98.0% (N)
Physical Form Crystalline Powder
Chemical Name or Material 1-(tert-Butoxycarbonyl)-4-[(p-toluenesulfonyloxy)methyl]piperidine