accessibility menu, dialog, popup

UserName

CAS RN 166815-96-9

IUPAC Name: tert-butyl 4-{[(4-methylbenzenesulfonyl)oxy]methyl}piperidine-1-carboxylate
Synonyms: tert-butyl 4-{[(4-methylbenzenesulfonyl)oxy]methyl}piperidine-1-carboxylate
Molecular Weight (g/mol): 369.48
Molecular Formula: C18H27NO5S
InChi Key: DARTVAOOTJKHQW-UHFFFAOYSA-N
SMILES: CC1=CC=C(C=C1)S(=O)(=O)OCC1CCN(CC1)C(=O)OC(C)(C)C
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results