missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-(tert-Butoxycarbonyl)-4-(methanesulfonyloxy)piperidine 98.0+%, TCI America™
$98.79 - $484.65
Chemical Identifiers
| CAS | 141699-59-4 |
|---|---|
| Molecular Formula | C11H21NO5S |
| Molecular Weight (g/mol) | 279.351 |
| MDL Number | MFCD04116220 |
| InChI Key | WOEQSXAIPTXOPY-UHFFFAOYSA-N |
| Synonym | 1-(tert-Butoxycarbonyl)-4-piperidyl Methanesulfonate, 1-Boc-4-(methanesulfonyloxy)piperidine, tert-Butyl 4-(Methanesulfonyloxy)piperidine-1-carboxylate, 4-(Methanesulfonyloxy)piperidine-1-carboxylic Acid tert-Butyl Ester |
| PubChem CID | 4184869 |
| IUPAC Name | tert-butyl 4-methylsulfonyloxypiperidine-1-carboxylate |
| SMILES | CC(C)(C)OC(=O)N1CCC(CC1)OS(=O)(=O)C |
Chemical Identifiers
| 141699-59-4 | |
| 279.351 | |
| WOEQSXAIPTXOPY-UHFFFAOYSA-N | |
| 4184869 | |
| CC(C)(C)OC(=O)N1CCC(CC1)OS(=O)(=O)C |
| C11H21NO5S | |
| MFCD04116220 | |
| 1-(tert-Butoxycarbonyl)-4-piperidyl Methanesulfonate, 1-Boc-4-(methanesulfonyloxy)piperidine, tert-Butyl 4-(Methanesulfonyloxy)piperidine-1-carboxylate, 4-(Methanesulfonyloxy)piperidine-1-carboxylic Acid tert-Butyl Ester | |
| tert-butyl 4-methylsulfonyloxypiperidine-1-carboxylate |
Specifications
| 141699-59-4 | |
| White | |
| MFCD04116220 | |
| 1-(tert-Butoxycarbonyl)-4-piperidyl Methanesulfonate, 1-Boc-4-(methanesulfonyloxy)piperidine, tert-Butyl 4-(Methanesulfonyloxy)piperidine-1-carboxylate, 4-(Methanesulfonyloxy)piperidine-1-carboxylic Acid tert-Butyl Ester | |
| CC(C)(C)OC(=O)N1CCC(CC1)OS(=O)(=O)C | |
| 279.351 | |
| 279.35 | |
| Crystalline Powder |
| 95°C | |
| C11H21NO5S | |
| 5 g | |
| WOEQSXAIPTXOPY-UHFFFAOYSA-N | |
| tert-butyl 4-methylsulfonyloxypiperidine-1-carboxylate | |
| 4184869 | |
| ≥98.0% (GC) | |
| 1-(tert-Butoxycarbonyl)-4-(methanesulfonyloxy)piperidine |
Safety and Handling
TSCA : No