accessibility menu, dialog, popup

UserName

CH3H3CCH3OONOSOOCH3

CAS RN 141699-59-4

IUPAC Name: tert-butyl 4-(methanesulfonyloxy)piperidine-1-carboxylate
Synonyms: tert-butyl 4-(methanesulfonyloxy)piperidine-1-carboxylate
Molecular Weight (g/mol): 279.35
Molecular Formula: C11H21NO5S
InChi Key: WOEQSXAIPTXOPY-UHFFFAOYSA-N
SMILES: CC(C)(C)OC(=O)N1CCC(CC1)OS(C)(=O)=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results