missing translation for 'onlineSavingsMsg'
Learn More

1-tert-Butoxycarbonyl-4-cyanopiperidine 97.0+%, TCI America™

Catalog No. B39621G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B39621G 1 g
B39625G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B39621G Supplier TCI America Supplier No. B39621G

Chemical Identifiers

CAS 91419-52-2
Molecular Formula C11H18N2O2
Molecular Weight (g/mol) 210.277
MDL Number MFCD01861223
InChI Key UQADQTBQNVARAP-UHFFFAOYSA-N
Synonym 1-boc-4-cyanopiperidine, 1-n-boc-4-cyanopiperidine, n-boc-4-cyanopiperidine, 1-n-boc-4-cyano-piperidine, n-boc-4-cyano-piperidine, 1-piperidinecarboxylic acid, 4-cyano-, 1,1-dimethylethyl ester, boc-4-cyanopiperidine, 4-cyano-piperidine-1-carboxylic acid tert-butyl ester, tert-butyl 4-cyano-1-piperidinecarboxylate, n-boc-piperidine-4-carbonitrile
PubChem CID 1514443
IUPAC Name tert-butyl 4-cyanopiperidine-1-carboxylate
SMILES CC(C)(C)OC(=O)N1CCC(CC1)C#N

Specifications

Melting Point 62°C
Color White
Quantity 1 g
UN Number 3439
Formula Weight 210.28
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1-tert-Butoxycarbonyl-4-cyanopiperidine
TSCA No
Recommended Storage Refrigerator