missing translation for 'onlineSavingsMsg'
Learn More

1-(tert-Butoxycarbonyl)-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyrazole 98.0+%, TCI America™

Catalog No. B52031G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B52031G 1 g
B52035G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B52031G Supplier TCI America Supplier No. B52031G

Chemical Identifiers

CAS 552846-17-0
Molecular Formula C14H23BN2O4
Molecular Weight (g/mol) 294.16
MDL Number MFCD05663873
InChI Key IPISOFJLWYBCAV-UHFFFAOYSA-N
Synonym 1-boc-pyrazole-4-boronic acid pinacol ester, tert-butyl 4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-1h-pyrazole-1-carboxylate, 1-boc-4-pyrazoleboronic acid pinacol ester, 1-boc-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyrazole, tert-butyl 4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyrazole-1-carboxylate, n-boc-pyrazole-4-boronic acid pinacol ester, 1-tert-butoxycarbonyl-1h-pyrazole-4-boronic acid, pinacol ester, n-boc-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-pyrazole, 1-tert-butoxycarbonyl-1h-pyrazol-4-yl boronic acid pinacol ester, 1-tert-butoxycarbonyl-4-1h-pyrazoleboronic acid
PubChem CID 16217654
IUPAC Name tert-butyl 4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazole-1-carboxylate
SMILES CC(C)(C)OC(=O)N1C=C(C=N1)B1OC(C)(C)C(C)(C)O1

Specifications

Melting Point 86°C
Color White
Quantity 1 g
Formula Weight 294.16
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 1-(tert-Butoxycarbonyl)-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyrazole
TSCA No
Recommended Storage Refrigerator