accessibility menu, dialog, popup

UserName

CAS RN 552846-17-0

IUPAC Name: tert-butyl 4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazole-1-carboxylate
Synonyms: tert-butyl 4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyrazole-1-carboxylate
Molecular Weight (g/mol): 294.16
Molecular Formula: C14H23BN2O4
InChi Key: IPISOFJLWYBCAV-UHFFFAOYSA-N
SMILES: CC(C)(C)OC(=O)N1C=C(C=N1)B1OC(C)(C)C(C)(C)O1
Percent Purity 
  • (2)
  • (2)
Physical Form 
  • (4)
Quantity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (4)
  • (2)
  • (2)