missing translation for 'onlineSavingsMsg'
Learn More

1-(tert-Butoxycarbonyl)-3-iodoazetidine 95.0+%, TCI America™

Catalog No. p-7123447
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B42881G 1 g
B42885G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B42881G Supplier TCI America Supplier No. B42881G
Only null left

Chemical Identifiers

CAS 254454-54-1
Molecular Formula C8H14INO2
Molecular Weight (g/mol) 283.11
MDL Number MFCD09752821
InChI Key XPDIKRMPZNLBAC-UHFFFAOYSA-N
Synonym 1-boc-3-iodoazetidine, 1-boc-3-iodo-azetidine, 1-boc-3-iodo azetidine, n-boc-3-iodoazetidine, 3-iodo-azetidine-1-carboxylic acid tert-butyl ester, 3-iodoazetidine, n-boc protected, 1-n-boc-3-iodoazetidine, 1-tert-butoxycarbonyl-3-iodoazetidine, tert-butyl 3-iodo-1-azetidinecarboxylate, tert-butyl 3-iodoazetidinecarboxylate
PubChem CID 11000522
IUPAC Name tert-butyl 3-iodoazetidine-1-carboxylate
SMILES CC(C)(C)OC(=O)N1CC(I)C1

Specifications

Color Yellow
Quantity 1 g
Formula Weight 283.11
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material 1-(tert-Butoxycarbonyl)-3-iodoazetidine