accessibility menu, dialog, popup

UserName

CAS RN 254454-54-1

IUPAC Name: tert-butyl 3-iodoazetidine-1-carboxylate
Synonyms: tert-butyl 3-iodoazetidine-1-carboxylate
Molecular Weight (g/mol): 283.11
Molecular Formula: C8H14INO2
InChi Key: XPDIKRMPZNLBAC-UHFFFAOYSA-N
SMILES: CC(C)(C)OC(=O)N1CC(I)C1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results