missing translation for 'onlineSavingsMsg'
Learn More

1-(tert-Butoxycarbonyl)-3-(hydroxymethyl)piperazine 98.0+%, TCI America™

Catalog No. B4716200MG
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B4716200MG 200 mg
B47161G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B4716200MG Supplier TCI America Supplier No. B4716200MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 301673-16-5
Molecular Formula C10H20N2O3
Molecular Weight (g/mol) 216.281
MDL Number MFCD04115304
InChI Key NSILYQWHARROMG-UHFFFAOYSA-N
Synonym 1-Boc-3-(hydroxymethyl)piperazine, tert-Butyl 3-(Hydroxymethyl)piperazine-1-carboxylate, 3-(Hydroxymethyl)piperazine-1-carboxylic Acid tert-Butyl Ester
PubChem CID 22386508
IUPAC Name tert-butyl 3-(hydroxymethyl)piperazine-1-carboxylate
SMILES CC(C)(C)OC(=O)N1CCNC(C1)CO

Specifications

Melting Point 95°C
Color White-Yellow
Quantity 200 mg
Formula Weight 216.28
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 1-(tert-Butoxycarbonyl)-3-(hydroxymethyl)piperazine