accessibility menu, dialog, popup

UserName

CAS RN 301673-16-5

IUPAC Name: tert-butyl 3-(hydroxymethyl)piperazine-1-carboxylate
Synonyms: tert-butyl 3-(hydroxymethyl)piperazine-1-carboxylate
Molecular Weight (g/mol): 216.28
Molecular Formula: C10H20N2O3
InChi Key: NSILYQWHARROMG-UHFFFAOYNA-N
SMILES: CC(C)(C)OC(=O)N1CCNC(CO)C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results