missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-(tert-Butoxycarbonyl)-3-azetidinone 98.0+%, TCI America™
$80.85 - $236.80
Chemical Identifiers
| CAS | 398489-26-4 |
|---|---|
| Molecular Formula | C8H13NO3 |
| Molecular Weight (g/mol) | 171.20 |
| MDL Number | MFCD01861741 |
| InChI Key | VMKIXWAFFVLJCK-UHFFFAOYSA-N |
| Synonym | 1-boc-3-azetidinone, 1-boc-azetidin-3-one, 1-n-boc-3-azetidinone, n-boc-3-azetidinone, n-boc-3-oxoazetidine, 1-tert-butoxycarbonyl-3-azetidinone, tert-butyl-3-oxoazetidine-1-carboxylate, 1-boc-3-oxoazetidine, n-boc-azetidin-3-one, 3-oxo-azetidine-1-carboxylic acid tert-butyl ester |
| PubChem CID | 1519404 |
| IUPAC Name | tert-butyl 3-oxoazetidine-1-carboxylate |
| SMILES | CC(C)(C)OC(=O)N1CC(=O)C1 |
Chemical Identifiers
| 398489-26-4 | |
| 171.20 | |
| VMKIXWAFFVLJCK-UHFFFAOYSA-N | |
| 1519404 | |
| CC(C)(C)OC(=O)N1CC(=O)C1 |
| C8H13NO3 | |
| MFCD01861741 | |
| 1-boc-3-azetidinone, 1-boc-azetidin-3-one, 1-n-boc-3-azetidinone, n-boc-3-azetidinone, n-boc-3-oxoazetidine, 1-tert-butoxycarbonyl-3-azetidinone, tert-butyl-3-oxoazetidine-1-carboxylate, 1-boc-3-oxoazetidine, n-boc-azetidin-3-one, 3-oxo-azetidine-1-carboxylic acid tert-butyl ester | |
| tert-butyl 3-oxoazetidine-1-carboxylate |
Specifications
| 398489-26-4 | |
| C8H13NO3 | |
| 1 g | |
| VMKIXWAFFVLJCK-UHFFFAOYSA-N | |
| tert-butyl 3-oxoazetidine-1-carboxylate | |
| 1519404 | |
| ≥98.0% (GC) | |
| 1-(tert-Butoxycarbonyl)-3-azetidinone |
| 52°C | |
| MFCD01861741 | |
| 1-boc-3-azetidinone, 1-boc-azetidin-3-one, 1-n-boc-3-azetidinone, n-boc-3-azetidinone, n-boc-3-oxoazetidine, 1-tert-butoxycarbonyl-3-azetidinone, tert-butyl-3-oxoazetidine-1-carboxylate, 1-boc-3-oxoazetidine, n-boc-azetidin-3-one, 3-oxo-azetidine-1-carboxylic acid tert-butyl ester | |
| CC(C)(C)OC(=O)N1CC(=O)C1 | |
| 171.20 | |
| 171.20 | |
| Crystalline Powder |
Safety and Handling
TSCA : No