missing translation for 'onlineSavingsMsg'
Learn More

1-(tert-Butoxycarbonyl)-3-azetidinone 98.0+%, TCI America™

Catalog No. b39885g
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
1 g
5 g
Catalog No. Quantity
B39885G 5 g
B39881G 1 g
2 options

Catalog No. B39885G

Supplier: TCI America B39885G

Only null left

Chemical Identifiers

CAS 398489-26-4
Molecular Formula C8H13NO3
Molecular Weight (g/mol) 171.20
MDL Number MFCD01861741
InChI Key VMKIXWAFFVLJCK-UHFFFAOYSA-N
Synonym 1-boc-3-azetidinone, 1-boc-azetidin-3-one, 1-n-boc-3-azetidinone, n-boc-3-azetidinone, n-boc-3-oxoazetidine, 1-tert-butoxycarbonyl-3-azetidinone, tert-butyl-3-oxoazetidine-1-carboxylate, 1-boc-3-oxoazetidine, n-boc-azetidin-3-one, 3-oxo-azetidine-1-carboxylic acid tert-butyl ester
PubChem CID 1519404
IUPAC Name tert-butyl 3-oxoazetidine-1-carboxylate
SMILES CC(C)(C)OC(=O)N1CC(=O)C1

Specifications

Melting Point 52°C
Quantity 5 g
Formula Weight 171.20
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1-(tert-Butoxycarbonyl)-3-azetidinone