missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-(tert-Butoxycarbonyl)-2-piperidinecarboxylic Acid 98.0+%, TCI America™
$67.38 - $240.64
Chemical Identifiers
| CAS | 98303-20-9 |
|---|---|
| Molecular Formula | C11H19NO4 |
| Molecular Weight (g/mol) | 229.276 |
| MDL Number | MFCD01862877 |
| InChI Key | JQAOHGMPAAWWQO-UHFFFAOYSA-N |
| Synonym | n-boc-2-piperidinecarboxylic acid, 1-tert-butoxycarbonyl piperidine-2-carboxylic acid, 1-boc-piperidine-2-carboxylic acid, n-boc-dl-pipecolinic acid, n-boc-piperidine-2-carboxylic acid, boc-dl-pip-oh, n-boc-pipecolic acid, 1-n-boc-2-piperidinecarboxylic acid, 1-tert-butoxy carbonyl piperidine-2-carboxylic acid, 1-tert-butoxycarbonyl-2-piperidinecarboxylic acid |
| PubChem CID | 581831 |
| IUPAC Name | 1-[(2-methylpropan-2-yl)oxycarbonyl]piperidine-2-carboxylic acid |
| SMILES | CC(C)(C)OC(=O)N1CCCCC1C(=O)O |
Chemical Identifiers
| 98303-20-9 | |
| 229.276 | |
| JQAOHGMPAAWWQO-UHFFFAOYSA-N | |
| 581831 | |
| CC(C)(C)OC(=O)N1CCCCC1C(=O)O |
| C11H19NO4 | |
| MFCD01862877 | |
| n-boc-2-piperidinecarboxylic acid, 1-tert-butoxycarbonyl piperidine-2-carboxylic acid, 1-boc-piperidine-2-carboxylic acid, n-boc-dl-pipecolinic acid, n-boc-piperidine-2-carboxylic acid, boc-dl-pip-oh, n-boc-pipecolic acid, 1-n-boc-2-piperidinecarboxylic acid, 1-tert-butoxy carbonyl piperidine-2-carboxylic acid, 1-tert-butoxycarbonyl-2-piperidinecarboxylic acid | |
| 1-[(2-methylpropan-2-yl)oxycarbonyl]piperidine-2-carboxylic acid |
Specifications
| 98303-20-9 | |
| White-Yellow | |
| MFCD01862877 | |
| n-boc-2-piperidinecarboxylic acid, 1-tert-butoxycarbonyl piperidine-2-carboxylic acid, 1-boc-piperidine-2-carboxylic acid, n-boc-dl-pipecolinic acid, n-boc-piperidine-2-carboxylic acid, boc-dl-pip-oh, n-boc-pipecolic acid, 1-n-boc-2-piperidinecarboxylic acid, 1-tert-butoxy carbonyl piperidine-2-carboxylic acid, 1-tert-butoxycarbonyl-2-piperidinecarboxylic acid | |
| CC(C)(C)OC(=O)N1CCCCC1C(=O)O | |
| 229.276 | |
| 229.28 | |
| Crystalline Powder |
| 131°C | |
| C11H19NO4 | |
| 5 g | |
| JQAOHGMPAAWWQO-UHFFFAOYSA-N | |
| 1-[(2-methylpropan-2-yl)oxycarbonyl]piperidine-2-carboxylic acid | |
| 581831 | |
| ≥98.0% (GC,T) | |
| 1-(tert-Butoxycarbonyl)-2-piperidinecarboxylic Acid |
Safety and Handling
TSCA : No