missing translation for 'onlineSavingsMsg'
Learn More

1-(tert-Butoxycarbonyl)-2-piperidinecarboxylic Acid 98.0+%, TCI America™

Catalog No. B36445G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B36445G 5 g
B364425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B36445G Supplier TCI America Supplier No. B36445G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 98303-20-9
Molecular Formula C11H19NO4
Molecular Weight (g/mol) 229.276
MDL Number MFCD01862877
InChI Key JQAOHGMPAAWWQO-UHFFFAOYSA-N
Synonym n-boc-2-piperidinecarboxylic acid, 1-tert-butoxycarbonyl piperidine-2-carboxylic acid, 1-boc-piperidine-2-carboxylic acid, n-boc-dl-pipecolinic acid, n-boc-piperidine-2-carboxylic acid, boc-dl-pip-oh, n-boc-pipecolic acid, 1-n-boc-2-piperidinecarboxylic acid, 1-tert-butoxy carbonyl piperidine-2-carboxylic acid, 1-tert-butoxycarbonyl-2-piperidinecarboxylic acid
PubChem CID 581831
IUPAC Name 1-[(2-methylpropan-2-yl)oxycarbonyl]piperidine-2-carboxylic acid
SMILES CC(C)(C)OC(=O)N1CCCCC1C(=O)O

Specifications

Melting Point 131°C
Color White-Yellow
Quantity 5 g
Formula Weight 229.28
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 1-(tert-Butoxycarbonyl)-2-piperidinecarboxylic Acid