missing translation for 'onlineSavingsMsg'
Learn More

1-tert-Butoxycarbonyl-1,2,3,6-tetrahydropyridine 97.0+%, TCI America™

Catalog No. B43571G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B43571G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B43571G Supplier TCI America Supplier No. B43571G

Chemical Identifiers

CAS 85838-94-4
Molecular Formula C10H17NO2
Molecular Weight (g/mol) 183.251
MDL Number MFCD04972245
InChI Key SHHHRQFHCPINIB-UHFFFAOYSA-N
Synonym n-boc-1,2,3,6-tetrahydropyridine, tert-butyl 5,6-dihydropyridine-1 2h-carboxylate, tert-butyl 1,2,3,6-tetrahydropyridine-1-carboxylate, tert-butyl 3,6-dihydropyridine-1 2h-carboxylate, 1-boc-1,2,3,6-tetrahydro-pyridine, 1-boc-1,2,3,6-tetrahydropyridine, n-boc-1,2,5,6-tetrahydropyridine, n-tert-butoxycarbonyl-1,2,3,6-tetrahydropyridine, 1 2h-pyridinecarboxylic acid, 3,6-dihydro-, 1,1-dimethylethyl ester, 3,6-dihydro-2h-pyridine-1-carboxylic acid tert-butyl ester
PubChem CID 13094787
IUPAC Name tert-butyl 3,6-dihydro-2H-pyridine-1-carboxylate
SMILES CC(C)(C)OC(=O)N1CCC=CC1

Specifications

Boiling Point 84°C
Color Yellow
Quantity 1 g
Formula Weight 183.25
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 1-tert-Butoxycarbonyl-1,2,3,6-tetrahydropyridine