missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ 1-Step™ Ultra TMB-Blotting Solution

Catalog No. PI37574
Change view
Click to view available options
Quantity:
250 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PI37574 250 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PI37574 Supplier Thermo Scientific™ Supplier No. 37574

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Thermo Scientific™ 1-Step Ultra TMB-Blotting Solution is an enhanced single-component horseradish peroxidase (HRP) substrate for Western blotting and immunohistochemistry.

Thermo Scientific™ 1-Step Ultra TMB-Blotting Solution is an enhanced single-component horseradish peroxidase (HRP) substrate for Western blotting and immunohistochemistry.

Additional Information and Online Ordering

Thermo Scientific 1-Step Ultra TMB-Blotting Solution is an enhanced single-component horseradish peroxidase (HRP) substrate for Western blotting.

This precipitating, colorimetric Western blot substrate for HRP provides high sensitivity, increased signal-to-noise ratio and low background compared to many other chromogenic substrates. The blotting solution contains soluble TMB (3,3',5,5'-tetramethylbenzidine), which reacts very quickly with horseradish peroxidase enzyme to produce an insoluble dark blue precipitate that does not fade or flake. The substrate is compatible with both nitrocellulose and PVDF membranes. The blotting solution is supplied ready to use with no mixing required.

Features of 1-Step Ultra TMB-Blotting Solution:
• Fast—protein bands visible in less than one minute
• Fade-resistant—protein bands stable after membrane drying
• Sensitive—detection limit similar to Pierce ECL Western Blotting Substrate
• Chromogenic—no special equipment needed for visualization; produces dark blue bands
• Ready to use—no organic solvents are required to dissolve; no dilution necessary for use

Applications:
• Chromogenic Western blot detection
• Nitrocellulose membrane blots
• PVDF membrane blots

Specifications

Product Type Substrate
Detection Method Colorimetric
Form Liquid
Membrane Compatibility Nitrocellulose, PVDF
Product Line 1-Step
Quantity 250 mL
Substrate TMB
Content And Storage Upon receipt store product at 4°C. Product is shipped at ambient temperature.

For Research Use Only. Not for use in diagnostic procedures.