missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ 1-Step™ TMB-Blotting Substrate Solution

Catalog No. pi34018
Change view
Click to view available options
Quantity:
250 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PI34018 250 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PI34018 Supplier Thermo Scientific™ Supplier No. 34018
Only null left

Ready-to-use blue TMB peroxidase (HRP) substrate specially formulated for chromogenic detection in Western blot and IHC experiments.

Thermo Scientific™ Pierce TMB-Blotting 1-Step Solution is a ready-to-use solution of the blue TMB peroxidase (HRP) substrate that is specially formulated for chromogenic detection in Western blotting and immunohistochemistry methods.

TMB (3,3',5,5'-tetramethylbenzidine; MW = 240.4) is most often used as a soluble substrate to detect the activity of peroxidase-conjugated antibodies in ELISA. Pierce TMB-Blotting Solution is formulated to precipitate and localize the blue reaction product onto membrane surfaces or tissue samples at sites where the peroxidase-substrate reaction occurs. TMB-Blotting Substrate produces a very large signal-to-noise ratio, although its high sensitivity requires careful washing and blocking to avoid background staining. When performing immunohistochemical techniques, aqueous counterstains and mounting medium must be used.

Highlights:

  • HRP substrate – for sensitive detection of horseradish peroxidase activity on solid media

  • Chromogenic – no special equipment needed for visualization; produces dark blue bands in Western blots; is not as susceptible to fading as AEC or 4-CN substrates

  • Ready-to-use – no organic solvents are required to dissolve; no dilution necessary for use

  • Complete – does not require other buffers or addition of hydrogen peroxide

Specifications

Product Type Substrate
Detection Method Colorimetric
Form Liquid
Product Line 1-Step
Quantity 250 mL
Substrate Horseradish Peroxidase
Reactivity Horseradish Peroxidase
Content And Storage Upon receipt store product at 4°C. Product is shipped at ambient temperature.

For Research Use Only. Not for use in diagnostic procedures.