missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ 1-Step™ Chloronaphthol Substrate Solution

Catalog No. FER34012
Change view
Click to view available options
Quantity:
250 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FER34012 250 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FER34012 Supplier Thermo Scientific™ Supplier No. 34012

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

1-Step™ Chloronaphthol Substrate Solution

Thermo Scientific Pierce 1-Step Chloronaphthol Solution is a solution of 4-chloro-1-naphthol for the chromogenic detection of horseradish peroxidase (HRP) activity in Western blot and tissue staining methods.

Chloronaphthol (4-CN, MW = 178.6) reacts with HRP to form a distinct blue to blue-purple product to localize and visualize the activity of peroxidase-conjugated antibodies on blots or fixed tissue samples. The HRP substrate is not as sensitive or stable as other chromogenic substrates, such as TMB and DAB. However, the blue-purple precipitate photographs well, and its alcohol-solubility is a property that can be utilized in certain double-staining applications.

Features of Chloronaphthol Substrates:

  • HRP substrate—for detection of horseradish peroxidase activity on solid media
  • Chromogenic—no special equipment needed for visualization
  • Blue-purple—easy to photograph in color or black-and-white
  • Package options—choose powder or ready-to-use solution

    Highlights of Pierce CN Solution
  • Ready-to-use—no ethanol or methanol required to dissolve; no dilution necessary for use
  • Complete—does not require buffer or addition of hydrogen peroxide
  • Pre-filtered—does not need to be filtered before use like powders or tablets after they are dissolved
  • Stable—store refrigerated at 4°C for at least one year

  • Specifications

    Product Type Substrate
    Detection Method Colorimetric
    Form Liquid
    Product Line 1-Step
    Quantity 250 mL
    Substrate Horseradish Peroxidase
    Reactivity Horseradish Peroxidase
    Content And Storage Store at 4°C.

    For Research Use Only. Not for use in diagnostic procedures.