missing translation for 'onlineSavingsMsg'
Learn More

1-sec-Butyl-2-nitrobenzene 98.0+%, TCI America™

Catalog No. B24275G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B24275G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B24275G Supplier TCI America Supplier No. B24275G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 19370-34-4
Molecular Formula C10H13NO2
Molecular Weight (g/mol) 179.219
InChI Key GMQQDASXGTUDPJ-UHFFFAOYSA-N
PubChem CID 86858
IUPAC Name 1-butan-2-yl-2-nitrobenzene
SMILES CCC(C)C1=CC=CC=C1[N+](=O)[O-]

Specifications

Boiling Point 126°C
Quantity 5 g
Formula Weight 179.22
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 1-sec-Butyl-2-nitrobenzene