missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Pyrrolidino-1-cyclopentene 97.0+%, TCI America™
Supplier: TCI America P100125G
Specifications
| 1-Pyrrolidino-1-cyclopentene | |
| Yellow | |
| C9H15N | |
| 25 g | |
| 1-(1-Cyclopenten-1-yl)pyrrolidine | |
| C1CCN(C1)C2=CCCC2 | |
| 137.23 | |
| 137.23 | |
| Liquid |
| 7148-07-4 | |
| 93°C | |
| MFCD00003162 | |
| 1993 | |
| KOFSFYBXUYHNJL-UHFFFAOYSA-N | |
| 1-(cyclopenten-1-yl)pyrrolidine | |
| 81570 | |
| ≥97.0% (GC) |
Chemical Identifiers
| 7148-07-4 | |
| 137.23 | |
| KOFSFYBXUYHNJL-UHFFFAOYSA-N | |
| 81570 | |
| C1CCN(C1)C2=CCCC2 |
| C9H15N | |
| MFCD00003162 | |
| 1-(1-Cyclopenten-1-yl)pyrrolidine | |
| 1-(cyclopenten-1-yl)pyrrolidine |
Safety and Handling
TSCA : Yes
Recommended Storage : Refrigerator